Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MMP25
Entrez Gene ID
64386
Gene Name
matrix metallopeptidase 25
Gene Aliases
MMP-25, MMP20, MMP20A, MMPL1, MT-MMP 6, MT-MMP6, MT6-MMP, MT6MMP, MTMMP6
Chromosome Location
Chr.16: 3045963 - 3060729 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022468.4 NP_071913.1 1 - 2 338 59
XM_017023561.1 XP_016879050.1 1 - 2 1056 59
GenBank mRNA AB042328.1 - 1 - 2 270 59
AF145442.2 - 1 - 2 125 59
AF185270.1 - 1 - 2 125 59
AJ239053.1 - 1 - 2 282 59
AJ272137.1 - 1 - 2 338 59

View Full Product Details