The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080433.1 | NP_001073902.1 | 4 - 5 | 1953 | 137 |
GenBank mRNA | AB067499.2 | - | 4 - 5 | 2037 | 137 |
BC131557.1 | - | 4 - 5 | 625 | 137 |