The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267562.1 | NP_001254491.1 | 1 - 2 | 348 | 63 |
NM_001881.3 | NP_001872.3 | 1 - 2 | 348 | 63 | |
NR_051975.1 | 1 - 2 | 348 | 63 | ||
XM_017015716.1 | XP_016871205.1 | 1 - 2 | 297 | 63 | |
XM_017015717.1 | XP_016871206.1 | 1 - 2 | 224 | 63 | |
XM_017015719.1 | XP_016871208.1 | 1 - 2 | 178 | 63 | |
XM_017015721.1 | XP_016871210.1 | 1 - 2 | 167 | 63 | |
XM_017015722.1 | XP_016871211.1 | 1 - 2 | 80 | 63 | |
XM_017015723.1 | XP_016871212.1 | 1 - 2 | 237 | 63 | |
XM_017015724.1 | XP_016871213.1 | 1 - 2 | 348 | 63 | |
XM_017015726.1 | XP_016871215.1 | 1 - 2 | 105 | 63 | |
XM_017015728.1 | XP_016871217.1 | 1 - 2 | 348 | 63 | |
XM_017015734.1 | XP_016871223.1 | 1 - 2 | 165 | 63 | |
GenBank mRNA | BC017117.1 | - | 1 - 2 | 212 | 63 |
BC066342.1 | - | 1 - 2 | 266 | 63 | |
BG573680.1 | - | 1 - 2 | 317 | 63 | |
BI459432.1 | - | 1 - 2 | 126 | 63 | |
DA756152.1 | - | 1 - 2 | 350 | 63 | |
DR760671.1 | - | 1 - 2 | 213 | 63 |