The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193369.1 | NP_001180298.1 | 11 - 12 | 2879 | 131 |
NM_001193370.1 | NP_001180299.1 | 11 - 12 | 2879 | 131 | |
NM_033081.2 | NP_149072.2 | 11 - 12 | 2923 | 131 | |
NM_080797.3 | NP_542987.2 | 11 - 12 | 2923 | 131 | |
XM_011528504.1 | XP_011526806.1 | 12 - 13 | 2964 | 131 | |
XM_011528505.1 | XP_011526807.1 | 12 - 13 | 3031 | 131 | |
XM_011528506.1 | XP_011526808.1 | 12 - 13 | 9815 | 131 | |
XM_011528507.1 | XP_011526809.1 | 12 - 13 | 2899 | 131 | |
XM_011528508.1 | XP_011526810.1 | 12 - 13 | 3040 | 131 | |
XM_011528509.2 | XP_011526811.1 | 11 - 12 | 2856 | 131 | |
GenBank mRNA | AB002331.2 | - | 11 - 12 | 2855 | 131 |
AY481571.1 | - | 11 - 12 | 2831 | 131 | |
AY481572.1 | - | 11 - 12 | 2831 | 131 | |
BC137177.1 | - | 10 - 11 | 2778 | 131 |