The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193369.1 | NP_001180298.1 | 8 - 9 | 2526 | 148 |
NM_001193370.1 | NP_001180299.1 | 8 - 9 | 2526 | 148 | |
NM_033081.2 | NP_149072.2 | 8 - 9 | 2570 | 148 | |
NM_080797.3 | NP_542987.2 | 8 - 9 | 2570 | 148 | |
XM_011528504.1 | XP_011526806.1 | 9 - 10 | 2611 | 148 | |
XM_011528505.1 | XP_011526807.1 | 9 - 10 | 2678 | 148 | |
XM_011528506.1 | XP_011526808.1 | 9 - 10 | 9462 | 148 | |
XM_011528507.1 | XP_011526809.1 | 9 - 10 | 2546 | 148 | |
XM_011528508.1 | XP_011526810.1 | 9 - 10 | 2687 | 148 | |
XM_011528509.2 | XP_011526811.1 | 8 - 9 | 2503 | 148 | |
GenBank mRNA | AB002331.2 | - | 8 - 9 | 2502 | 148 |
AY481571.1 | - | 8 - 9 | 2478 | 148 | |
AY481572.1 | - | 8 - 9 | 2478 | 148 | |
BC137177.1 | - | 7 - 8 | 2425 | 148 |