The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037763.2 | NP_001032852.2 | 13 - 14 | 1286 | 166 |
XM_011515358.2 | XP_011513660.1 | 13 - 14 | 6131 | 166 | |
XM_011515359.2 | XP_011513661.1 | 13 - 14 | 1289 | 166 | |
XM_011515360.2 | XP_011513662.1 | 13 - 14 | 2571 | 166 | |
XM_011515363.2 | XP_011513665.1 | 13 - 14 | 2574 | 166 | |
XM_011515364.2 | XP_011513666.1 | 13 - 14 | 2573 | 166 | |
XM_011515365.2 | XP_011513667.1 | 13 - 14 | 2574 | 166 | |
XM_011515366.2 | XP_011513668.1 | 13 - 14 | 2574 | 166 | |
XM_017012131.1 | XP_016867620.1 | 14 - 15 | 7307 | 166 | |
XM_017012132.1 | XP_016867621.1 | 13 - 14 | 2571 | 166 | |
XM_017012133.1 | XP_016867622.1 | 13 - 14 | 2575 | 166 | |
GenBank mRNA | AJ890451.1 | - | 13 - 14 | 1286 | 166 |
AJ890453.1 | - | 8 - 9 | 561 | 166 | |
BC136892.1 | - | 13 - 14 | 1276 | 166 |