The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037763.2 | NP_001032852.2 | 16 - 17 | 1488 | 118 |
XM_011515358.2 | XP_011513660.1 | 16 - 17 | 6333 | 118 | |
XM_011515359.2 | XP_011513661.1 | 16 - 17 | 1491 | 118 | |
XM_011515360.2 | XP_011513662.1 | 16 - 17 | 2773 | 118 | |
XM_011515362.2 | XP_011513664.1 | 4 - 5 | 293 | 118 | |
XM_011515363.2 | XP_011513665.1 | 16 - 17 | 2776 | 118 | |
XM_011515364.2 | XP_011513666.1 | 16 - 17 | 2775 | 118 | |
XM_011515365.2 | XP_011513667.1 | 16 - 17 | 2776 | 118 | |
XM_011515366.2 | XP_011513668.1 | 16 - 17 | 2776 | 118 | |
XM_017012131.1 | XP_016867620.1 | 17 - 18 | 7509 | 118 | |
XM_017012132.1 | XP_016867621.1 | 16 - 17 | 2773 | 118 | |
XM_017012133.1 | XP_016867622.1 | 16 - 17 | 2777 | 118 | |
GenBank mRNA | AJ890451.1 | - | 16 - 17 | 1488 | 118 |
AJ890453.1 | - | 11 - 12 | 763 | 118 | |
BC136892.1 | - | 16 - 17 | 1478 | 118 | |
BC144431.1 | - | 17 - 18 | 1663 | 118 |