Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL28A1
Entrez Gene ID
340267
Gene Name
collagen type XXVIII alpha 1 chain
Gene Aliases
COL28
Chromosome Location
Chr.7: 7338543 - 7542062 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037763.2 NP_001032852.2 19 - 20 1699 148
XM_011515358.2 XP_011513660.1 19 - 20 6544 148
XM_011515359.2 XP_011513661.1 19 - 20 1702 148
XM_011515360.2 XP_011513662.1 19 - 20 2984 148
XM_011515362.2 XP_011513664.1 7 - 8 504 148
XM_011515363.2 XP_011513665.1 19 - 20 2987 148
XM_011515364.2 XP_011513666.1 19 - 20 2986 148
XM_011515365.2 XP_011513667.1 19 - 20 2987 148
XM_011515366.2 XP_011513668.1 19 - 20 2987 148
XM_017012131.1 XP_016867620.1 20 - 21 7720 148
XM_017012132.1 XP_016867621.1 19 - 20 2984 148
XM_017012133.1 XP_016867622.1 19 - 20 2988 148
GenBank mRNA AJ890451.1 - 19 - 20 1699 148
AJ890453.1 - 14 - 15 974 148
BC136892.1 - 19 - 20 1689 148
BC144431.1 - 20 - 21 1874 148

View Full Product Details