Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IGLV3-25
Entrez Gene ID
28793
Gene Name
immunoglobulin lambda variable 3-25
Gene Aliases
IGLV325, V2-17
Chromosome Location
Chr.22: 22686769 - 22687281 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC022823.1 - 1 - 2 81 116
BC089418.1 - 1 - 2 56 116

View Full Product Details