Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRC1
Entrez Gene ID
9055
Gene Name
protein regulator of cytokinesis 1
Gene Aliases
ASE1
Chromosome Location
Chr.15: 90966038 - 90994651 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267580.1 NP_001254509.1 12 - 13 1784 124
XM_005254987.2 XP_005255044.1 13 - 14 1815 124
XM_017022717.1 XP_016878206.1 12 - 13 1692 124
GenBank mRNA AK297117.1 - 12 - 13 1658 124
BG823292.1 - 3 - 4 259 124

View Full Product Details