Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PRC1
- Entrez Gene ID
-
9055
- Gene Name
- protein regulator of cytokinesis 1
- Gene Aliases
-
ASE1
- Chromosome Location
-
Chr.15: 90966038 - 90994651 on Build GRCh38
View Full Product Details