The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_194449.3 | NP_919431.2 | 7 - 8 | 2888 | 90 |
XM_011525886.1 | XP_011524188.1 | 7 - 8 | 2882 | 90 | |
GenBank mRNA | AB011178.2 | - | 7 - 8 | 1601 | 90 |
AK001924.1 | - | 7 - 8 | 1162 | 90 | |
BC014927.2 | - | 7 - 8 | 1175 | 90 | |
BC082244.1 | - | 2 - 3 | 315 | 90 | |
BC126277.1 | - | 7 - 8 | 1223 | 90 |