The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173564.3 | NP_775835.2 | 2 - 3 | 227 | 155 |
XM_006715907.3 | XP_006715970.1 | 2 - 3 | 282 | 155 | |
XM_017011868.1 | XP_016867357.1 | 2 - 3 | 282 | 155 | |
GenBank mRNA | AK124773.1 | - | 2 - 3 | 227 | 155 |