Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
FOXP3 | NM_001114377.1 | NP_001107849.1 | ||
NM_014009.3 | NP_054728.2 | |||
XM_006724533.2 | XP_006724596.2 | |||
XM_011543916.2 | XP_011542218.1 | |||
XM_017029565.1 | XP_016885054.1 | |||
XM_017029566.1 | XP_016885055.1 | |||
XM_017029567.1 | XP_016885056.1 | |||
AF277993.1 | AAG53607.1 | |||
AJ005891.1 | CAA06748.1 | |||
AK292052.1 | ||||
AK299988.1 | ||||
BC113401.1 | ||||
BC113403.1 | ||||
BC143785.1 | ||||
BC143786.1 | ||||
DQ010327.1 | AAY27088.1 | |||
EF534714.1 | ||||
EU855812.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv471602 | ChrX:49205142 - 49320261 on Build GRCh38 | Gain | PPP1R3F GAGE10 CCDC22 CACNA1F FOXP3 |
nsv528292 | ChrX:48433286 - 49274368 on Build GRCh38 | Gain | MAGIX PRAF2 PORCN CCDC22 RBM3 TBC1D25 SLC35A2 TFE3 WAS HDAC6 PIM2 FTSJ1 SLC38A5 GLOD5 EBP SYP-AS1 SYP PQBP1 GATA1 OTUD5 PRICKLE3 CACNA1F FOXP3 SUV39H1 KCND1 PCSK1N PPP1R3F PLP2 CCDC120 WDR13 LOC101927635 TIMM17B ERAS GRIPAP1 WDR45 GPKOW |
esv32909 | ChrX:48003374 - 49285646 on Build GRCh38 | Gain+Loss | MAGIX PRAF2 PORCN CCDC22 RBM3 SSX1 TBC1D25 SLC35A2 TFE3 WAS HDAC6 PIM2 FTSJ1 SLC38A5 GLOD5 EBP SYP-AS1 SSX3 SYP SSX6 PQBP1 SPACA5B ZNF182 GATA1 SSX4B OTUD5 PRICKLE3 CACNA1F SSX9 FOXP3 SUV39H1 KCND1 SPACA5 PCSK1N PPP1R3F PLP2 CCDC120 WDR13 SSX4 LOC101927635 ZNF630-AS1 TIMM17B ERAS ZNF630 GRIPAP1 WDR45 GPKOW SSX5 |
nsv469734 | ChrX:49205141 - 49320261 on Build GRCh38 | Gain+Loss | PPP1R3F GAGE10 CCDC22 CACNA1F FOXP3 |
nsv6895 | ChrX:49227319 - 49262690 on Build GRCh38 | Insertion | CCDC22 CACNA1F FOXP3 |