Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CFAP54
Entrez Gene ID
144535
Gene Name
cilia and flagella associated protein 54
Gene Aliases
C12orf55, C12orf63
Chromosome Location
Chr.12: 96489566 - 96875555 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306084.1 NP_001293013.1 47 - 48 6565 131
XM_011539069.2 XP_011537371.1 48 - 49 6765 131
XM_011539070.2 XP_011537372.1 48 - 49 6765 131
XM_011539071.2 XP_011537373.1 48 - 49 6765 131
XM_011539072.2 XP_011537374.1 48 - 49 6765 131
XM_011539073.2 XP_011537375.1 48 - 49 6765 131
XM_011539074.2 XP_011537376.1 48 - 49 6765 131
XM_011539076.2 XP_011537378.1 48 - 49 6765 131
XM_011539077.2 XP_011537379.1 47 - 48 6915 131
XM_011539078.2 XP_011537380.1 48 - 49 6765 131
XM_011539079.1 XP_011537381.1 44 - 45 6011 131
XM_011539081.2 XP_011537383.1 48 - 49 6765 131
XM_011539083.2 XP_011537385.1 48 - 49 6765 131
XM_011539084.2 XP_011537386.1 48 - 49 6765 131
XM_011539085.1 XP_011537387.1 33 - 34 4586 131
XM_017018866.1 XP_016874355.1 48 - 49 6765 131
XM_017018867.1 XP_016874356.1 37 - 38 5017 131
GenBank mRNA AK126100.1 - 14 - 15 1879 131

View Full Product Details