The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198923.2 | NP_944605.2 | 1 - 1 | 860 | 94 |
GenBank mRNA | AB154410.1 | - | 1 - 1 | 860 | 94 |
AY427820.1 | - | 1 - 1 | 860 | 94 |