The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145719.1 | NP_001139191.1 | 2 - 3 | 276 | 58 |
NM_207372.2 | NP_997255.2 | 2 - 3 | 775 | 58 | |
XM_017016236.1 | XP_016871725.1 | 2 - 3 | 3190 | 58 | |
GenBank mRNA | AK123885.1 | - | 2 - 3 | 775 | 58 |
AK123978.1 | - | 2 - 3 | 276 | 58 |