The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011513603.2 | XP_011511905.1 | 8 - 9 | 3266 | 73 |
XM_011513604.2 | XP_011511906.1 | 9 - 10 | 3532 | 73 | |
XM_017008891.1 | XP_016864380.1 | 10 - 11 | 4065 | 73 | |
XM_017008892.1 | XP_016864381.1 | 10 - 11 | 4065 | 73 | |
XM_017008893.1 | XP_016864382.1 | 9 - 10 | 3898 | 73 | |
GenBank mRNA | AK128339.1 | - | 6 - 7 | 670 | 73 |
BC140710.1 | - | 6 - 7 | 646 | 73 |