Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LGALS17A
Entrez Gene ID
400696
Gene Name
galectin 14 pseudogene
Gene Aliases
-
Chromosome Location
Chr.19: 39679374 - 39686373 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_034156.1 1 - 2 67 76
GenBank mRNA AK123499.1 - 1 - 2 67 76
FJ613354.1 - 1 - 2 48 76

View Full Product Details