Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBE2NL
Entrez Gene ID
389898
Gene Name
ubiquitin conjugating enzyme E2 N like (gene/pseudogene)
Gene Aliases
Li174
Chromosome Location
Chr.X: 143884071 - 143885255 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012989.2 NP_001013007.1 1 - 1 53 140
NR_121210.1 1 - 1 53 140
GenBank mRNA GU727624.1 - 1 - 1 53 140

View Full Product Details