The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001524.1 | NP_001515.1 | 2 - 2 | 520 | 105 |
GenBank mRNA | AF041240.1 | - | 2 - 2 | 520 | 105 |