Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
KANSL1 | NM_001193465.1 | NP_001180394.1 | ||
NM_001193466.1 | NP_001180395.1 | |||
NM_015443.3 | NP_056258.1 | |||
XM_006721823.1 | XP_006721886.1 | |||
XM_006721824.3 | XP_006721887.1 | |||
XM_011524628.2 | XP_011522930.1 | |||
XM_011524629.2 | XP_011522931.1 | |||
XM_011524630.2 | XP_011522932.1 | |||
XM_011524631.2 | XP_011522933.1 | |||
XM_011524632.2 | XP_011522934.1 | |||
XM_017024488.1 | XP_016879977.1 | |||
AB033093.1 | BAA86581.1 | |||
AK094946.1 | ||||
AK291259.1 | ||||
AK310569.1 | 1 | 866 | ||
AL117476.1 | CAB55949.1 | |||
AL137317.1 | CAB70694.1 | |||
BC035892.1 | AAH35892.1 | |||
BC098376.1 | AAH98376.1 | |||
BX538006.1 | CAD97958.1 | |||
BX648760.1 | CAH10565.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv471698 | Chr17:46002113 - 46300887 on Build GRCh38 | Gain+Loss | MAPT ARL17B KANSL1-AS1 KANSL1 LRRC37A |
nsv469705 | Chr17:45969304 - 46127121 on Build GRCh38 | Loss | MAPT KANSL1 STH |
nsv469537 | Chr17:46002112 - 46163353 on Build GRCh38 | Gain+Loss | MAPT KANSL1 |
nsv575142 | Chr17:46003698 - 46172742 on Build GRCh38 | Loss | MAPT KANSL1 |
esv2762433 | Chr17:45496768 - 46710828 on Build GRCh38 | Gain+Loss | SPPL2C MGC57346-CRHR1 LOC107987253 CRHR1-IT1 KANSL1-AS1 ARL17A MAPT-IT1 NSFP1 KANSL1 LRRC37A MGC57346 MAPT-AS1 LOC644172 LOC105376839 NSF CRHR1 MAPT ARL17B LOC107987254 LRRC37A2 LRRC37A4P STH |
nsv1072740 | Chr17:45970433 - 46248034 on Build GRCh38 | Deletion | MAPT KANSL1-AS1 KANSL1 STH |
dgv3178n100 | Chr17:45877004 - 46034461 on Build GRCh38 | Gain | MAPT MAPT-IT1 KANSL1 MAPT-AS1 STH |
esv2751686 | Chr17:45962198 - 46286690 on Build GRCh38 | Gain | MAPT ARL17B KANSL1-AS1 KANSL1 LRRC37A STH |
esv2758692 | Chr17:46006548 - 47199967 on Build GRCh38 | Gain+Loss | RPS2P47 WNT9B LOC107987253 KANSL1-AS1 ARL17A NSFP1 KANSL1 LRRC37A GOSR2 NSF MIR5089 MAPT ARL17B LOC107987254 LRRC37A2 WNT3 RPRML CDC27 |
dgv128e55 | Chr17:46017105 - 46286690 on Build GRCh38 | Gain | MAPT ARL17B KANSL1-AS1 KANSL1 LRRC37A |
dgv127e55 | Chr17:45927606 - 46733750 on Build GRCh38 | Gain | NSF LOC107987253 MAPT ARL17B LOC107987254 KANSL1-AS1 LRRC37A2 ARL17A NSFP1 KANSL1 LRRC37A STH |