The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005473.2 | NP_001005473.1 | 2 - 3 | 911 | 148 |
XM_017009436.1 | XP_016864925.1 | 2 - 3 | 833 | 148 | |
XM_017009437.1 | XP_016864926.1 | 2 - 3 | 1631 | 148 | |
XM_017009438.1 | XP_016864927.1 | 2 - 3 | 939 | 148 | |
GenBank mRNA | AK127142.1 | - | 2 - 3 | 884 | 148 |
BX648329.1 | - | 2 - 3 | 912 | 148 |