The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002125.3 | NP_002116.2 | 5 - 6 | 792 | 143 |
GenBank mRNA | BC009234.2 | - | 5 - 6 | 830 | 143 |
M15992.1 | - | 4 - 5 | 700 | 143 | |
M16086.1 | - | 4 - 5 | 570 | 143 | |
M20429.1 | - | 5 - 6 | 792 | 143 | |
M32578.1 | - | 5 - 6 | 848 | 143 | |
M57648.1 | - | 5 - 6 | 845 | 143 |