The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024533.4 | NP_078809.2 | 3 - 3 | 2781 | 84 |
GenBank mRNA | AF246718.1 | - | 3 - 3 | 2782 | 84 |