Product Details

Assay Reference Genome
Location

Chr.X:107650780 on build GRCh38
Cytoband
Xq22.3
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
5631
Gene Symbol
PRPS1
Gene Name
phosphoribosyl pyrophosphate synthetase 1
Gene Aliases
ARTS, CMTX5, DFN2, DFNX1, PPRibP, PRS-I, PRSI
Location
Chr.X:107628424-107651026 on build GRCh38
Assay Gene Location
Within Exon 7
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
PRPS1 NM_001204402.1 4 1502 NP_001191331.1
NM_002764.3 7 1910 NP_002755.1
AK316467.1 4 1416
BC001605.1 7 1805 AAH01605.1
D00860.1 7 1827 BAA00733.1

View Full Product Details