Product Details
- Assay Reference Genome
Location
Chr.Y:12726595 on build GRCh38
- Cytoband
- Yq11.221
- Species
- Homo sapiens
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 8287
- Gene Symbol
- USP9Y
- Gene Name
- ubiquitin specific peptidase 9, Y-linked
- Gene Aliases
- DFFRY, SPGFY2
- Location
- Chr.Y:12701231-12860844 on build GRCh38
- Assay Gene Location
- Overlaps Intron 7 - Exon 8
View Full Product Details