Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NETO1
Entrez Gene ID
81832
Gene Name
neuropilin and tolloid like 1
Gene Aliases
BCTL1, BTCL1
Chromosome Location
Chr.18: 72742314 - 72867667 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138999.1 NP_620552.1 5 - 5 1150 76
XM_017026024.1 XP_016881513.1 5 - 5 1330 76
GenBank mRNA AF448839.1 - 5 - 5 1150 76

View Full Product Details