The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010922.2 | NP_001010922.1 | 1 - 2 | 300 | 73 |
GenBank mRNA | AI821052.1 | - | 1 - 2 | 300 | 73 |
AK289439.1 | - | 1 - 2 | 334 | 73 | |
AY265861.1 | - | 1 - 2 | 128 | 73 | |
AY265862.1 | - | 1 - 2 | 128 | 73 | |
BC127719.1 | - | 1 - 2 | 161 | 73 | |
BC127720.1 | - | 1 - 2 | 161 | 73 |