The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195290.1 | NP_001182219.1 | 3 - 4 | 255 | 100 |
NM_205841.3 | NP_995313.2 | 2 - 3 | 382 | 100 | |
GenBank mRNA | AY358716.1 | - | 2 - 3 | 102 | 100 |
BC032003.1 | - | 2 - 3 | 335 | 100 | |
BE122852.1 | - | 2 - 3 | 171 | 100 | |
GQ504704.1 | - | 2 - 3 | 400 | 100 | |
GQ504705.1 | - | 2 - 3 | 336 | 100 |