The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039905.2 | NP_001034994.1 | 1 - 2 | 246 | 98 |
GenBank mRNA | AK095915.1 | - | 1 - 2 | 246 | 98 |