The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077191.1 | NP_001070659.1 | 2 - 2 | 1755 | 146 |
NM_001077194.1 | NP_001070662.1 | 2 - 2 | 1247 | 146 | |
NM_001321950.1 | NP_001308879.1 | 2 - 2 | 1621 | 146 | |
NM_170699.2 | NP_733800.1 | 2 - 2 | 1134 | 146 | |
XM_011510743.1 | XP_011509045.1 | 2 - 2 | 2242 | 146 | |
XM_017003467.1 | XP_016858956.1 | 3 - 3 | 2359 | 146 | |
XM_017003468.1 | XP_016858957.1 | 3 - 3 | 2980 | 146 | |
XM_017003469.1 | XP_016858958.1 | 3 - 3 | 2472 | 146 | |
GenBank mRNA | AB086170.1 | - | 1 - 1 | 887 | 146 |
AK122658.1 | - | 2 - 2 | 1621 | 146 | |
AK122660.1 | - | 2 - 2 | 1755 | 146 | |
BC033625.1 | - | 2 - 2 | 1118 | 146 |