The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243007.1 | NP_001229936.1 | 2 - 3 | 1417 | 77 |
XM_005267543.3 | XP_005267600.1 | 5 - 6 | 2495 | 77 | |
GenBank mRNA | BC105720.1 | - | 2 - 3 | 1416 | 77 |
BC105927.1 | - | 2 - 3 | 1416 | 77 |