The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004536.2 | NP_004527.2 | 14 - 15 | 4317 | 146 |
NM_022892.1 | NP_075043.1 | 12 - 13 | 3646 | 146 | |
XM_011543413.2 | XP_011541715.1 | 4 - 5 | 2455 | 146 | |
XM_017009490.1 | XP_016864979.1 | 4 - 5 | 3002 | 146 | |
GenBank mRNA | AB048534.1 | - | 12 - 13 | 3646 | 146 |
AB209123.1 | - | 3 - 4 | 3703 | 146 | |
AK124511.1 | - | 3 - 4 | 2998 | 146 | |
AK295115.1 | - | 3 - 4 | 1545 | 146 | |
BC136273.1 | - | 12 - 13 | 3654 | 146 | |
BC143761.1 | - | 11 - 12 | 3486 | 146 | |
U19251.1 | - | 14 - 15 | 3890 | 146 |