The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004536.2 | NP_004527.2 | 15 - 16 | 4401 | 96 |
NM_022892.1 | NP_075043.1 | 13 - 14 | 3730 | 96 | |
XM_011543413.2 | XP_011541715.1 | 5 - 6 | 2539 | 96 | |
XM_017009490.1 | XP_016864979.1 | 5 - 6 | 3086 | 96 | |
GenBank mRNA | AB048534.1 | - | 13 - 14 | 3730 | 96 |
AB209123.1 | - | 4 - 5 | 3787 | 96 | |
AK124511.1 | - | 4 - 5 | 3082 | 96 | |
AK295115.1 | - | 4 - 5 | 1629 | 96 | |
BC136273.1 | - | 13 - 14 | 3738 | 96 | |
BC143761.1 | - | 12 - 13 | 3570 | 96 | |
U19251.1 | - | 15 - 16 | 3974 | 96 |