Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MARCH11
Entrez Gene ID
441061
Gene Name
membrane associated ring-CH-type finger 11
Gene Aliases
MARCH-XI
Chromosome Location
Chr.5: 16067365 - 16179788 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102562.1 NP_001096032.1 1 - 2 748 82

View Full Product Details