The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243961.1 | NP_001230890.1 | 1 - 2 | 192 | 57 |
NM_002123.4 | NP_002114.3 | 1 - 2 | 192 | 57 | |
GenBank mRNA | AB209580.1 | - | 1 - 2 | 141 | 57 |
AK316333.1 | - | 1 - 2 | 184 | 57 | |
DC409119.1 | - | 1 - 2 | 192 | 57 | |
M20432.1 | - | 1 - 2 | 169 | 57 | |
M25327.1 | - | 1 - 2 | 187 | 57 |