Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- HCG26
- Entrez Gene ID
-
352961
- Gene Name
- HLA complex group 26 (non-protein coding)
- Gene Aliases
-
3.8-1, NCRNA00191, bCX205D4.4, bPG181B23.4
- Chromosome Location
-
Chr.6: 31471229 - 31472408 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_002812.3 |
|
1 - 1 |
1057 |
110 |
GenBank mRNA |
L29376.1 |
- |
1 - 1 |
1056 |
111 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BQ185253.1 |
- |
1 |
|
|
Chr. 6 - 31471949 - 31472410 [-] on Build GRCh38 |
110 |
GenBankmRNA |
AA534999.1 |
- |
1 |
|
|
Chr. 6 - 31471995 - 31472410 [-] on Build GRCh38 |
110 |
GenBankmRNA |
CB046175.1 |
- |
1 |
|
|
Chr. 6 - 31471935 - 31472410 [-] on Build GRCh38 |
109 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
HLA complex P5 (non-protein coding) |
HCP5 |
6S2650E, D6S2650E, P5-1 |
-5.4kb
|
Sense |
Chr. - 31463180 - 31465809 on Build GRCh38 |
Downstream |
uncharacterized LOC102725068 |
LOC102725068 |
|
7.5kb
|
Anti-sense |
Chr. - 31479918 - 31494794 on Build GRCh38 |
View Full Product Details