The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_001459.2 | 1 - 1 | 102 | 148 | |
NR_003138.3 | 1 - 1 | 102 | 148 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | small Cajal body-specific RNA 13 | SCARNA13 | U93 | -64.0 | Sense | Chr. - 95533355 - 95533629 on Build GRCh38 |
Downstream | glutaredoxin 5 | GLRX5 | C14orf87, FLB4739, GRX5, PR01238, PRO1238, PRSA, SIDBA3, SPAHGC | 114.0 | Anti-sense | Chr. - 95534986 - 95544718 on Build GRCh38 |