Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- NSAP11
- Entrez Gene ID
-
100131275
- Gene Name
- nervous system abundant protein 11
- Gene Aliases
-
-
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AY176665.1 |
- |
1 - 2 |
295 |
81 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC105379305 |
LOC105379305 |
|
-108.8kb
|
Anti-sense |
Chr. - 18608999 - 18612069 on Build GRCh38 |
Downstream |
uncharacterized LOC100128993 |
LOC100128993 |
|
453.3kb
|
Anti-sense |
Chr. - 19183674 - 19245522 on Build GRCh38 |
View Full Product Details