The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
GenBank mRNA | BC044607.1 | - | 1 - 1 | 955 | 70 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | small nucleolar RNA host gene 16 | SNHG16 | ncRAN | -4.4kb | Sense | Chr. - 76557764 - 76565348 on Build GRCh38 |
Downstream | ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 1 | ST6GALNAC1 | HSY11339, SIAT7A, ST6GalNAcI, STYI | 46.5kb | Anti-sense | Chr. - 76617769 - 76643838 on Build GRCh38 |