Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- FYB
- Entrez Gene ID
-
2533
- Gene Name
- FYN binding protein
- Gene Aliases
-
ADAP, PRO0823, SLAP-130, SLAP130
- Chromosome Location
-
Chr.5: 39105252 - 39274523 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BQ227053.1 |
- |
1 |
578 |
|
Chr. 5 - 39137521 - 39138351 [-] on Build GRCh38 |
93 |
GenBankmRNA |
H57330.1 |
- |
1 |
|
|
Chr. 5 - 39137704 - 39138142 [+] on Build GRCh38 |
93 |
GenBankmRNA |
DB327868.1 |
- |
1 |
|
|
Chr. 5 - 39137404 - 39137931 [+] on Build GRCh38 |
93 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
RPTOR independent companion of MTOR complex 2 |
RICTOR |
AVO3, PIA, hAVO3 |
-63.0kb
|
Sense |
Chr. - 38937920 - 39074421 on Build GRCh38 |
Downstream |
complement component 9 |
C9 |
ARMD15, C9D |
144.2kb
|
Sense |
Chr. - 39284275 - 39364553 on Build GRCh38 |
View Full Product Details