The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_026543.1 | 2 - 3 | 609 | 76 | |
GenBank mRNA | BC062296.1 | - | 2 - 3 | 611 | 76 |
BC080530.1 | - | 2 - 3 | 608 | 76 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | SH3 domain binding glutamate rich protein | SH3BGR | 21-GARP | -90.3kb | Anti-sense | Chr. - 39445865 - 39515506 on Build GRCh38 |
Downstream | beta-1,3-galactosyltransferase 5 | B3GALT5 | B3GalT-V, B3GalTx, B3T5, GLCT5, beta-1,3-GalTase 5, beta-3-Gx-T5, beta3Gal-T5 | 3.3kb | Anti-sense | Chr. - 39612940 - 39662889 on Build GRCh38 |