Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA U18302.1 - 1 249 Chr. 16 - 10901559 - 10902037 [+] on Build GRCh38 105


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream trans-golgi network vesicle protein 23 homolog A TVP23A FAM18A, YDR084C -82.7kb Anti-sense Chr. - 10758034 - 10818902 on Build GRCh38
Downstream Dexi homolog DEXI MYLE 26.9kb Anti-sense Chr. - 10928891 - 10943007 on Build GRCh38

View Full Product Details