Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- UROS
- Entrez Gene ID
-
7390
- Gene Name
- uroporphyrinogen III synthase
- Gene Aliases
-
UROIIIS
- Chromosome Location
-
Chr.10: 125784980 - 125823268 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC024936.1 |
- |
1 - 1 |
342 |
69 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BG028484.1 |
- |
1 |
341 |
|
Chr. 10 - 125791845 - 125792571 [-] on Build GRCh38 |
69 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
matrix metallopeptidase 21 |
MMP21 |
HTX7, MMP-21 |
-6.3kb
|
Sense |
Chr. - 125766453 - 125775821 on Build GRCh38 |
Downstream |
microRNA 4484 |
MIR4484 |
mir-4484 |
27.2kb
|
Anti-sense |
Chr. - 125819740 - 125819822 on Build GRCh38 |
View Full Product Details