Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BC040998.1 - 1 2049 Chr. 7 - 112478503 - 112480724 [-] on Build GRCh38 73


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream leucine rich single-pass membrane protein 1 LSMEM1 C7orf53 Anti-sense Chr. - 112480219 - 112491062 on Build GRCh38
Downstream uncharacterized LOC100996249 LOC100996249 137.3kb Anti-sense Chr. - 112618065 - 112620649 on Build GRCh38

View Full Product Details