Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- C9orf3
- Entrez Gene ID
-
84909
- Gene Name
- chromosome 9 open reading frame 3
- Gene Aliases
-
AOPEP, AP-O, APO, C90RF3, ONPEP
- Chromosome Location
-
Chr.9: 94726604 - 95148264 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AW469589.1 |
- |
1 |
|
|
Chr. 9 - 94874863 - 94875323 [-] on Build GRCh38 |
82 |
GenBankmRNA |
BE505030.1 |
- |
1 |
|
|
Chr. 9 - 94874792 - 94875320 [-] on Build GRCh38 |
82 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 2278 |
MIR2278 |
mir-2278 |
-62.3kb
|
Sense |
Chr. - 94809962 - 94810057 on Build GRCh38 |
Downstream |
uncharacterized LOC101928119 |
LOC101928119 |
|
45.0kb
|
Anti-sense |
Chr. - 94920339 - 94937841 on Build GRCh38 |
View Full Product Details