Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK123730.1 - 1 2045 Chr. 7 - 127992322 - 127994549 [+] on Build GRCh38 84
GenBankmRNA BU726320.1 - 1 Chr. 7 - 127994101 - 127994566 [-] on Build GRCh38 84
GenBankmRNA BF594070.1 - 1 Chr. 7 - 127994250 - 127994544 [-] on Build GRCh38 84
GenBankmRNA BG236464.1 - 1 Chr. 7 - 127994064 - 127994552 [-] on Build GRCh38 84
GenBankmRNA BE857387.1 - 1 Chr. 7 - 127994123 - 127994549 [-] on Build GRCh38 84
GenBankmRNA AW118739.1 - 1 Chr. 7 - 127994082 - 127994552 [-] on Build GRCh38 84
GenBankmRNA AW001186.1 - 1 Chr. 7 - 127993993 - 127994557 [-] on Build GRCh38 84
GenBankmRNA AW966387.1 - 1 314 Chr. 7 - 127994053 - 127994566 [+] on Build GRCh38 84
GenBankmRNA BU677687.1 - 1 Chr. 7 - 127993864 - 127994567 [-] on Build GRCh38 84
GenBankmRNA AI056975.1 - 1 Chr. 7 - 127994117 - 127994549 [-] on Build GRCh38 84
GenBankmRNA DB361431.1 - 1 Chr. 7 - 127994005 - 127994549 [-] on Build GRCh38 84
GenBankmRNA BM690994.1 - 1 432 Chr. 7 - 127993942 - 127994550 [+] on Build GRCh38 84
GenBankmRNA BQ448613.1 - 1 Chr. 7 - 127993910 - 127994567 [-] on Build GRCh38 84
GenBankmRNA AW303807.1 - 1 Chr. 7 - 127994151 - 127994549 [-] on Build GRCh38 84
GenBankmRNA DB319678.1 - 1 Chr. 7 - 127994012 - 127994549 [-] on Build GRCh38 84
GenBankmRNA DW424757.1 - 1 90 Chr. 7 - 127994294 - 127994563 [+] on Build GRCh38 84
GenBankmRNA AU253957.1 - 1 Chr. 7 - 127993880 - 127994565 [-] on Build GRCh38 84


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream staphylococcal nuclease and tudor domain containing 1 SND1 TDRD11, p100 Sense Chr. - 127652119 - 128092607 on Build GRCh38
Downstream SND1 intronic transcript 1 SND1-IT1 C7orf54, NAG8, NSG-X 3.0kb Sense Chr. - 127997509 - 128000077 on Build GRCh38

View Full Product Details