Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GSDMB
- Entrez Gene ID
-
55876
- Gene Name
- gasdermin B
- Gene Aliases
-
GSDML, PP4052, PRO2521
- Chromosome Location
-
Chr.17: 39904595 - 39919377 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK025174.1 |
- |
1 - 1 |
1252 |
104 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BQ303821.1 |
- |
1 |
570 |
|
Chr. 17 - 39916897 - 39917554 [-] on Build GRCh38 |
104 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
zona pellucida binding protein 2 |
ZPBP2 |
ZPBPL |
-38.4kb
|
Anti-sense |
Chr. - 39868164 - 39877896 on Build GRCh38 |
Downstream |
ORMDL sphingolipid biosynthesis regulator 3 |
ORMDL3 |
|
2.8kb
|
Sense |
Chr. - 39921041 - 39927804 on Build GRCh38 |
View Full Product Details