Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR1L6
Entrez Gene ID
392390
Gene Name
olfactory receptor family 1 subfamily L member 6
Gene Aliases
HG16, OR1K1, OR1L7, OR9-30
Chromosome Location
Chr.9: 122749848 - 122750783 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004453.2 NP_001004453.2 1 - 1 746 107

View Full Product Details